missing translation for 'onlineSavingsMsg'
Learn More

SUCNR1/GPR91 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP1-00861AF350

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213784

  • 623.09€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

SUCNR1/GPR91 Polyclonal specifically detects SUCNR1/GPR91 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

SUCNR1/GPR91
Polyclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
GPR91G protein-coupled receptor 91, G-protein coupled receptor 91, P2Y purinoceptor 1, P2Y purinoceptor 1-like, succinate receptor 1
Rabbit
0.1 mL
56670
Human, Mouse, Rat
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
SUCNR1
A synthetic peptide made to an internal portion of the human GPR91 protein (between residues 100-200) [Uniprot: Q9BXA5]
Primary
GPR91 (A135) detects endogenous levels of GPR91 protein.
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales