Learn More
Cole-Parmer™ Stuart™ Plastic Coated Receiving Flask
Product Code. 11982729
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
11982729 500 mL Each
1 options
This item is not returnable. View return policy
Product Code. 11982729 Supplier Cole-Parmer™ Supplier No. 1009673
  • ONLINE EXCLUSIVE PRICE
    260.00€
    220.06€ / paquete de 1
    Ahorro 39.94€
    15% Descuento
 Restricted Item
Fecha estimada de envoi: 14-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Material Borosilicate Glass
Type Receiving Flask
For Use With (Equipment) For RE100 Rotary Evaporator
Material (Coating) Plastic
Volume (Metric) 500 mL