Learn More
Corning™ Stripwell™ Microplates
Product Code. 10681983 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Clear
White
Binding Property:
High Binding
Medium Binding
Total Volume (Metric) Well:
190 μL
360 μL
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Binding Property Total Volume (Metric) Well Unit Size
10681983 White High Binding 190 μL Pack of 100
10347891 White Medium Binding 360 μL Pack of 100
10525531 Black Medium Binding 360 μL Pack of 100
10419331 Black High Binding 360 μL Pack of 100
10414872 Clear High Binding 190 μL Pack of 100
10517691 Clear High Binding 360 μL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10681983 Supplier Corning™ Supplier No. 2485
  • ONLINE EXCLUSIVE PRICE
    1035.00€
    964.65€ / paquete de 100
    Ahorro 70.35€
    6% Descuento
View all promotions
Fecha estimada de envoi: 09-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Order Info
Bulk packed. Cat. No. 07-200-24 has individual 1 x 8 strips without frame.
Certifications
Cat. Nos. 07-200-31 and -32 have certified surface chemistry.
TRUSTED_SUSTAINABILITY
Specifications
Binding Property High Binding
Format Strip Plate
Lid Without Lid
Array 8 x 12
Packaging 4 inner packs of 25
Binding Type High Bind
Color White
Product Type Microplate Strip
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 125 μL
Sterility Non-sterile
Surface Treatment High Binding
Total Volume (Metric) Well 190 μL
Well Shape Round
For Use With (Application) In-vitro diagnostic assays
No. per Case 100/Cs.
No. per Pack 25/Pk.
Quantity 100/Cs.