Learn More
IBA Lifesciences™ Strep-Tactin™ XT Coated Microplate
Product Code. 17812628 Shop All IBA Lifesciences Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Each
Product Code. Color Unit Size
17812628 Clear Each
1 options
This item is not returnable. View return policy
Product Code. 17812628 Supplier IBA Lifesciences™ Supplier No. 25101001
  • ONLINE EXCLUSIVE PRICE
    91.30€
    77.00€ / paquete de 1
    Ahorro 14.30€
    15% Descuento
View all promotions
 Restricted Item
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Ready-to-use 96-well microplate for assay applications
Due to the high specificity and affinity (nM-pM) of the streptavidin variant Strep-Tactin™ XT, Strep-tag™II or Twin-Strep-tag™, fusion proteins can be selectively and stably captured even from complex mixtures. Biomolecules of interest are presented to interacting partners in a uniform manner resulting in reliable and reproducible assays.
  • Ready-to-use Strep-Tactin™XT coated microplate (12 x 8-well strips) for ligand binding assays, immunoassays, enzyme assays, or diagnostic assays under high-throughput conditions.
  • The solid-phase, multi-well format is compatible with standard multichannel pipettes and automated plate readers.
  • Twin-Strep-tag™ fusion protein binding capacity determined with bacterial alkaline phosphatase (97 kDa) is 4 pmol for 200 μL incubation volume.
Specifications
Barcode No Barcode
Array 12 x 8
Autoclavable Not Autoclavable
Color Clear
Material Plastic
No. of Wells 96
Sterility Non-sterile
Surface Treatment Strep-TactinXT
Well Shape Flat Bottom (F-well)
For research use only.