missing translation for 'onlineSavingsMsg'
Learn More

IBA GmbH™ Strep-Tactin™ Superflow™ Cartridge

IBA GmbH™ Strep-Tactin™ Superflow™ Cartridge is a reliable one-step purification of Strep-tag™ fusion proteins with unsurpassed purity and bioacitivity.

Marca:  IBA GmbH™ 2-1235-001

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15331645

  • 88.00€ / 1 ml


Fecha de envío estimada
del proveedor 30-01-2019
Añadir a la cesta

Descripción

Descripción

  • For use with chromatography workstations with 10-32 connections (please note that you do not need adapters for HPLC and Äkta)
  • One step purification of recombinant Strep-tag™ proteins with HPLC or FPLC
  • They can also be operated with other workstations, with syringes or with peristaltic pumps by use of common adapters for cartridges
  • Cartridge bodies are pressure resistant for up to 3 bar, the cartridges can be used with a flow restrictor
  • Cartridges can be connected in series to enlarge capacity
  • Binding capacity: 50 to 100nmol recombinant Strep-tag™ protein (up to 3mg in case of a 30KDa protein) per 1mL bed volume
Especificación

Especificación

Chromatography workstations with 10 to 32 connections
Cartridge
1 x 1mL
Videos