missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17362-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Involved in spermatogenesis and sperm function. Plays a role in regulation of cell growth. Binds to double-stranded DNA and RNA. Binds most efficiently to poly(I:C) RNA than to poly(dI:dC) DNA. Binds also to single-stranded poly(G) RNA. Binds non-specifically to the mRNA PRM1 3′-UTR and adenovirus VA RNA.Especificaciones
STRBP | |
Polyclonal | |
Unconjugated | |
STRBP | |
ILF3L, p74, SPNR, STRBP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20744, 55342, 84476 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91WM1, Q96SI9, Q9JKU6 | |
STRBP | |
STRBP Fusion Protein Ag10720 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |