Learn More
Seward™ Stomacher™ 80 Biomaster Blender
Product Code. 12819405
Change view
Click to view available options
Volume Range (Metric):
5 to 80 mL
Unit Size:
Each
Product Code. Volume Range (Metric) Unit Size
12819405 5 to 80 mL Each
1 options
This item is not returnable. View return policy
Product Code. 12819405 Supplier Seward™ Supplier No. 0080/000/EU
  • ONLINE EXCLUSIVE PRICE
    4574.02€
    3119.47€ / paquete de 1
    Ahorro 1454.55€
    31% Descuento
Fecha estimada de envoi: 25-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The Stomacher™ Model 80 Biomaster provide small tissue processing solutions for clinical, life science and microbiology applications.
  • Capable of processing samples from 250μL to 80mL, the Biomaster caters for the increased demand for smaller samples without reducing sample quality
  • Allow samples to be processed safely and without risk of cross-contamination while maximizing visible cell recovery
  • Adjustable process times and paddle speeds
  • Proven enhancement in cell recovery and viability
  • Cost-effective sample preparation
Specifications
Voltage 230 V
Electrical Requirements 230 V, 50 Hz
Volume Range (Metric) 5 to 80 mL
Height (Metric) 190 mm
Width (Metric) 290 mm
For Use With (Application) For TB Work with Sputa and Tissues, Quality Control on Ointment and Cream Samples and Tissues in Animal Research
Frequency 50 Hz