Learn More
Sampling Systems™ SteriWare™ PP White Disposable PharmaScoop™
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ PP White Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
Marca: Sampling Systems™ 8045P-500S
Detalles adicionales : Peso : 61.92000kg
Descripción
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged in ultra-strong packing with easy to open design
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Large variety of different sizes available
- Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
- Positioned the ergonomic handle high up on the scoop
- No Cross Contamination
- No Cleaning required
Especificación
White | |
Individually Bagged | |
Sterile | |
Disposable Scoop |
Yes | |
PP (Polypropylene) | |
500mL |