Learn More
Sampling Systems™ SteriWare™ PP White Disposable PharmaScoop™
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ PP White Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
Marca:  Sampling Systems™ 8045P-500
Detalles adicionales : Peso : 61.92000kg
 Ver más versiones de este producto
Código de producto. 15879431
  • 324.60€ / paquete de 100
Fecha de envío estimada
del proveedor 08-03-2019
Añadir a la cesta
Descripción
Descripción
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged in ultra-strong packing with easy to open design
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Large variety of different sizes available
  • Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
  • Positioned the ergonomic handle high up on the scoop
  • No Cross Contamination
  • No Cleaning required
Especificación
Especificación
White
Individually Bagged
Non-Sterile
Disposable Scoop
Yes
PP (Polypropylene)
500mL
Videos