missing translation for 'onlineSavingsMsg'
Learn More

STAT5b Antibody (STAT5B/2611), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79907R

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213308

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

STAT5b
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
STAT5B
Recombinant full-length human STAT5b protein (Uniprot: P51692)
0.1 mL
Phospho Specific, Signal Transduction
Human
IgG1 κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
STAT5B/2611
Immunohistochemistry, Immunohistochemistry-Paraffin
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales