Learn More
Abnova™ STARD3 Recombinant Protein
Human STARD3 full-length ORF recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00010948-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of a subfamily of lipid trafficking proteins that are characterized by a C-terminal steroidogenic acute regulatory domain and an N-terminal metastatic lymph node 64 domain. The encoded protein localizes to the membranes of late endosomes and may be involved in exporting cholesterol. Alternate splicing results in multiple transcript variants.
- Theoretical MW (kDa): 74.69
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MSKLPRELTRDLERSLPAVASLGSSLSHSQSLSSHLLPPPEKRRAISDVRRTFCLFVTFDLLFISLLWIIELNTNTGIRKNLEQEIIQYNFKTSFFDIFVLAFFRFSGLLLGYAVLQLRHWWVIAVTTLVSSAFLIVKVILSELLSKGAFGYLLPIVSFVLAWLETWFLDFKVLPQEAEEERWYLAAQVAVARGPLLFSGALSEGQFYSPPESFAGSDNESDEEVAGKKSFSAQEREYIRQGKEATAVVDQILAQEENWKFEKNNEYGDTVYTIEVPFHGKTFILKTFLPCPAELVYQEVILQPERMVLWNKTVTACQILQRVEDNTLISYDVSAGAAGGVVSPRDFVNVRRIERRRDRYLSSGIATSHSAKPPTHKYVRGENGPGGFIVLKSASNPRVCTFVWILNTDLKGRLPRYLIHQSLAATMFEFAFHLRQRISELGARA
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH08747 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
74.69 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MSKLPRELTRDLERSLPAVASLGSSLSHSQSLSSHLLPPPEKRRAISDVRRTFCLFVTFDLLFISLLWIIELNTNTGIRKNLEQEIIQYNFKTSFFDIFVLAFFRFSGLLLGYAVLQLRHWWVIAVTTLVSSAFLIVKVILSELLSKGAFGYLLPIVSFVLAWLETWFLDFKVLPQEAEEERWYLAAQVAVARGPLLFSGALSEGQFYSPPESFAGSDNESDEEVAGKKSFSAQEREYIRQGKEATAVVDQILAQEENWKFEKNNEYGDTVYTIEVPFHGKTFILKTFLPCPAELVYQEVILQPERMVLWNKTVTACQILQRVEDNTLISYDVSAGAAGGVVSPRDFVNVRRIERRRDRYLSSGIATSHSAKPPTHKYVRGENGPGGFIVLKSASNPRVCTFVWILNTDLKGRLPRYLIHQSLAATMFEFAFHLRQRISELGARA | |
CAB1/FLJ41370/MLN64/es64 | |
STARD3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
10948 | |
STARD3 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
STARD3 | |
Human | |
Recombinant | |
Solution |