Learn More
STAR Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech CL594-12225-100UL
Detalles adicionales : Peso : 0.00010kg
Código de producto. 17254776
  • 430.00€ / 100 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock
Descripción
Descripción
Steroidogenic acute regulatory protein (StAR) is a key player in acute regulation of hormone-induced steroidogenesis, by facilitating cholesterol transport from cellular stores to inner mitochondria membrane where the cholesterol was conversed to pregnenolone. StAR is prevalently expressed in mitochondria of steroid-producing adrenal and gonadal tissue.
Especificaciones
Especificaciones
STAR
Polyclonal
CoraLite 594
100 μL
6770
Liquid
Immunofluorescence
1000 μg/mL