Learn More
Ismatec™ Standard Tubing Connectors, Plastic
Variously sized and shaped tubing connectors. Ismatec™ Standard Tubing Connectors, Plastic are available in different plastic compositions.
Marca:  Ismatec™ ISM513
Detalles adicionales : Peso : 0.10000kg
 Ver más versiones de este producto
Código de producto. 10216662
  • 70.65€ / paquete de 10
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Standard, non-reducing.
  • Designed for Ismatec™ pump tubing.
  • Available in different sizes, shapes, and compositions.
Especificaciones
Especificaciones
Acetal Resin (POM, Polyoxymethylene)
For 6.0mm I.D. Tubing
6.0mm O.D.
Standard Tubing Connector
10/Pk.
T-shape