Learn More
Cole-Parmer™ Essentials Utility Bowls, 304 Stainless Steel
Product Code. 11797255 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
858 mL
2000 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11797255 858 mL Each
11707265 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11797255 Supplier Cole-Parmer™ Essentials Supplier No. 0727200
  • ONLINE EXCLUSIVE PRICE
    53.00€
    40.47€ / paquete de 1
    Ahorro 12.53€
    23% Descuento
Fecha estimada de envoi: 22-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Heavy gauge, 304 stainless steel (SS) Utility Bowls with flat bottom and straight sides to ensure stability.
  • These straight-sided, heavy-duty bowls are made of 304 SS with a satin finish that makes them easy to clean.
Specifications
Material Stainless Steel
Capacity (Metric) 858 mL
Capacity (English) 29 oz.
Height (Metric) 6.7 cm
Diameter (English) 5 in.
Diameter (Metric) 127 mm
Height (English) 2.6 in.
Product Type Utility Bowl