Learn More
Aalborg™ Stainless Steel Support Frame for 150mm P-meter
Product Code. 11718378
Change view
Click to view available options
For Use With (Equipment):
Single Tube P-Meter
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11718378 Single Tube P-Meter Each
1 options
Product Code. 11718378 Supplier Aalborg™ Supplier No. P11S2VA0
  • ONLINE EXCLUSIVE PRICE
    332.00€
    169.32€ / paquete de 1
    Ahorro 162.68€
    49% Descuento
 Restricted Item
En stock
Add to basket
Edge
Add to basket
Aalborg™ Stainless Steel Support Frame for 150mm P-meter is for use with single tube flow meters, which are available with built in valves.
  • Rib-guided or fluted metering tubes facilitate stable and accurate readings
  • Magnifier lens in front shield to enhance reading resolution
  • Interchangeability of Flow Tubes and floats
  • Ease of installation and exchange of flow tubes
  • Glass flow tubes are prevented from turning during the tightening phase of the assembly procedure
  • Chemical compatibility
  • Simple means of panel mounting
  • Frame for P-meter
Specifications
For Use With (Equipment) Single Tube P-Meter
Type Support Frame
Width (Metric) 150 mm