Learn More
Bellingham Stanley™ Glass Polarimeter Tube
Product Code. 11960068
Change view
Click to view available options
Length (Metric):
100 mm
Unit Size:
Each
Product Code. Length (Metric) Unit Size
11960068 100 mm Each
1 options
This item is not returnable. View return policy
Product Code. 11960068 Supplier Bellingham Stanley™ Supplier No. 3510
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
  • Center Fill
  • Metal End Caps
TRUSTED_SUSTAINABILITY
Specifications
Type Polarimeter Tube
Material Stainless Steel, Glass
For Use With (Application) Aggressive chemicals and solvents
Diameter (Metric) 8 mm ID
Length (Metric) 100 mm
Wavelength 300 to 900 nm
Volume (Metric) 5 mL