Learn More
Bochem™ Stainless Steel Powder Funnel
Product Code. 10527371
Change view
Click to view available options
Diameter (Metric)Top:
240 mm
Unit Size:
Each
Product Code. Diameter (Metric)Top Unit Size
10527371 240 mm Each
1 options
This item is not returnable. View return policy
Product Code. 10527371 Supplier Bochem™ Supplier No. 8723
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specially designed to allow for the flow of powdered materials. Bochem™ Stainless Steel Powder Funnel is highly durable and resistant.
  • 18/10 stainless steel construction.
  • Straight and wide shaft.
Specifications
Diameter (Metric)Top 240 mm
Diameter (Metric) Stem 30mm
Material Stainless Steel (18/10)
Length (Metric) Stem 60mm
Length (Metric) Overall 210mm