Learn More
Bochem™ Stainless Steel Rollo Transport Cart
Product Code. 10562108
Change view
Click to view available options
Dimensions (L x W):
400 x 400 mm
Unit Size:
Each
Product Code. Dimensions (L x W) Unit Size
10562108 400 x 400 mm Each
1 options
This item is not returnable. View return policy
Product Code. 10562108 Supplier Bochem™ Supplier No. 8004
Fecha estimada de envoi: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Type Rollo transport cart
Load Bearing Capacity (Metric) 100 kg
Material Stainless Steel
Dimensions (L x W) 400 x 400 mm
Dimensions (L x W x H) 400 x 400 x 810 mm
Height (Metric) 810 mm
Length (Metric) 400 mm
Width (Metric) 400 mm