Learn More
Polvo de acero inoxidable, malla -325, tipo 316-L, Thermo Scientific Chemicals
Description
Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Nombre del producto químico o material | Stainless Steel powder, Type 316-L |
Color | Gris |
Peso Mol. medio o Intervalo de pesos mol. | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
Número MDL | MFCD00148499 |
Cantidad | 100 g |
Información de solubilidad | Insoluble in water. |
Tamaño de malla | −325 Mesh |
Formulario | Polvo |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.