Learn More
EVOS™ EVOS™ Stage Plate for Automated Stage Vessel Holders
EVOS™ vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides.
Marca:  EVOS™ AMEPVH048
Detalles adicionales : Peso : 0.50000kg
Código de producto. 15491074
  • 321.00€ / cada uno
Fecha estimada de envío
30-10-2020
Añadir a la cesta
Descripción
Descripción
Product Features:
  • EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment.
  • EVOS stage plates hold vessel holders, thereby expanding the number of vessel holders available for the EVOS systems.
  • With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Especificaciones
Especificaciones
EVOS XL/FL Stage Plate for EVOS Auto/OSI VH
220 mm x 220 mm
Room Temperature
Stage Plate for Automated Stage Vessel Holders
EVOS™ FL Auto Imaging System
EVOS™
EVOS™ FL Auto Imaging System