missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST3GAL5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14614-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Ganglioside GM3 is known to participate in the induction of cell differentiation, modulation of cell proliferation, maintece of fibroblast morphology, signal transduction, and integrin-mediated cell adhesion. The protein encoded by this gene is a type II membrane protein which catalyzes the formation of GM3 using lactosylceramide as the substrate. The encoded protein is a member of glycosyltransferase family 29 and may be localized to the Golgi apparatus. Mutation in this gene has been associated with Amish infantile epilepsy syndrome. Transcript variants encoding different isoforms have been found for this gene.Especificaciones
ST3GAL5 | |
Polyclonal | |
Unconjugated | |
ST3GAL5 | |
Ganglioside GM3 synthase, Sialyltransferase 9, SIAT9, SIATGM3S, ST3Gal V, ST3GAL5, ST3GalV | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20454, 83505, 8869 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88829, Q68G12, Q9UNP4 | |
ST3GAL5 | |
ST3GAL5 Fusion Protein Ag6207 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |