Learn More
GVS SPR534 Elipse Integra Mask with ABEK1P3 Filters
Click to view available options
:
GVS SPR534 with ABEK1P3 Filters
Descripción
This mask is recommended for multigas and dust risks, in chemical production and handling environment.
- Suitable all types of organic gases, inorganic gases, formaldehyde, chemical gases, dust and ammonia.
- Extremely lightweight with integrated eye and respirator protection to give a better face fit, more comfort, and reduce the risk of mist building up inside the lens.
- Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
- Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
Especificaciones
Especificaciones
Color | Azul |
Desechables | Reusable |