Learn More
GVS SPR534 Elipse Integra Mask with ABEK1P3 Filters
GREENER_CHOICE
Código de producto. 18459625
Click to view available options
:
GVS SPR534 with ABEK1P3 Filters
This mask is recommended for multigas and dust risks, in chemical production and handling environment. Suitable all types of organic gases, inorganic gases, formaldehyde, chemical gases, dust and ammonia.
This mask is recommended for multigas and dust risks, in chemical production and handling environment.
  • Suitable all types of organic gases, inorganic gases, formaldehyde, chemical gases, dust and ammonia.
  • Extremely lightweight with integrated eye and respirator protection to give a better face fit, more comfort, and reduce the risk of mist building up inside the lens.
  • Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
  • Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
Especificaciones
Color Azul
Desechables Reusable