Learn More
GVS SPR401 Elipse Integra Mask with A1P3 Filters for Organic Gases and Dust
Click to view available options
:
GVS SPR401 with A1P3 Filters
Descripción
This mask is recommended for organic gases and dust, pesticides, painting, solvents into automotive, construction, agriculture and shipyard industry or repair.
- Extremely lightweight with integrated eye and respirator protection to give a better face fit, more comfort, and reduce the risk of mist building up inside the lens.
- Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
Especificaciones
Especificaciones
Color | Azul |
Desechables | Reusable |