Learn More
Spinosad, TRC
Product Code. 30004718
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004718 50 mg 50mg
30012794 25 mg 25mg
30003637 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004718 Supplier Toronto Research Chemicals Supplier No. TRCS68192050MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 168316-95-8
Molecular Formula C42 H67 N O10 . C41 H65 N O10
Molecular Weight (g/mol) 747
Synonym Blackhawk, Comfortis, Conserve, Entrust, Extinosad, GF 120, GF 120 Fruit Fly Bait, GF 120 NF Naturalyte, GF 120 Naturalyte Fruit Fly Bait, GF 1587, Gardens Alive Bulls-Eye, LY 232105, Laser, Laser (insecticide), Naturalyte, PP 105, SpinTor, Spinoace, Spinosad NT, Spinosyn A-spinosyn D mixt., Success, Success 4, Success Naturalyte, Tracer, Tracer Naturalyte, Tracer Naturalyte Insect Control, XDE 105
IUPAC Name (1S,2S,5R,7S,9S,10S,14R,15S,19S)-15-[(2R,5S,6R)-5-(dimethylamino)-6-methyloxan-2-yl]oxy-19-ethyl-4,14-dimethyl-7-[(2R,3R,4R,5S,6S)-3,4,5-trimethoxy-6-methyloxan-2-yl]oxy-20-oxatetracyclo[10.10.0.02,10.05,9]docosa-3,11-diene-13,21-dione
SMILES CC[C@H]1CCC[C@H](O[C@H]2CC[C@@H]([C@@H](C)O2)N(C)C)[C@@H](C)C(=O)C3=C[C@H]4[C@@H]5C[C@@H](C[C@H]5C(=C[C@H]4[C@@H]3CC(=O)O1)C)O[C@@H]6O[C@@H](C)[C@H](OC)[C@@H](OC)[C@H]6OC
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C42H67NO10/c1-11-26-13-12-14-35(53-37-16-15-34(43(6)7)24(4)49-37)23(3)38(45)33-20-31-29(32(33)21-36(44)51-26)17-22(2)28-18-27(19-30(28)31)52-42-41(48-10)40(47-9)39(46-8)25(5)50-42/h17,20,23-32,34-35,37,39-42H,11-16,18-19,21H2,1-10H3/t23-,24-,25+,26+,27-,28+,29-,30-,31-,32+,34+,35+,37+,39+,40-,41-,42+/m1/s1
Formula Weight 745.4765
Percent Purity >95%
Chemical Name or Material Spinosad
Grade HPLC