missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPCS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11847-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.Especificaciones
SPCS1 | |
Polyclonal | |
Unconjugated | |
SPCS1 | |
HSPC033, SPase 12 kDa subunit, SPC1, SPC12, SPCS1, YJR010C A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28972 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y6A9 | |
SPCS1 | |
SPCS1 Fusion Protein Ag2407 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |