Learn More
Sparteine Sulfate, TRC
Product Code. 30011717
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011717 250 mg 250mg
30011274 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011717 Supplier Toronto Research Chemicals Supplier No. TRCS679500250MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 299-39-8
Molecular Formula C15 H26 N2 . H2 O4 S
Molecular Weight (g/mol) 332.46
Synonym 7,14-Methano-2H,6H-dipyrido[1,2-a:1',2'-e][1,5]diazocine, dodecahydro-, (7S,7aR,14S,14aS)-, sulfate (1:1), 7,14-Methano-2H,6H-dipyrido[1,2-a:1',2'-e][1,5]diazocine, dodecahydro-, [7S-(7α,7aα,14α,14aβ)]-, sulfate (1:1), Sparteine, sulfate (1:1) (8CI), (-)-Sparteine sulfate, NSC 143087, NSC 26253, NSC 402663, Spal, Spareng, Sparm, Sparteal, Sparteine sulfate, Spartepur, Spartin, Spartocin, Spatym, Tocosimplex
SMILES OS(=O)(=O)O.C1CCN2C[C@@H]3C[C@@H](CN4CCCC[C@@H]34)[C@H]2C1
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C15H26N2.H2O4S/c1-3-7-16-11-13-9-12(14(16)5-1)10-17-8-4-2-6-15(13)17;1-5(2,3)4/h12-15H,1-11H2;(H2,1,2,3,4)/t12-,13-,14-,15+;/m0./s1
Formula Weight 332.177
Chemical Name or Material Sparteine Sulfate