missing translation for 'onlineSavingsMsg'
Learn More

SPARC Antibody (001), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90612AF647

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213378

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

SPARC Monoclonal specifically detects SPARC in Mouse samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

SPARC
Monoclonal
Alexa Fluor 647
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified
Western Blot, ELISA
1
Western Blot, ELISA
Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin)
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SPARC (NP_033268.1; Met1-Ile302).
0.1 mL
Biologically Active Proteins, Cancer, Cell Cycle and Replication
6678
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales