missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPANXA2/SPANXA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19527-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
SPANXA1, also named as SPANXA and SPANXA2, is a member of the SPANX family of cancer/testis-associated genes. It encodes differentially expressed testis-specific proteins that localize to various subcellular compartments. On Western blots of human testis proteins, SPANX antibodies reacts with the bands migrating between 15–30 kDa. The SPANX protein may be a useful target in cancer immunotherapy.Especificaciones
SPANXA2/SPANXA1 | |
Polyclonal | |
Unconjugated | |
SPANXA2 | |
Cancer/testis antigen 11.1, CT11.1, SPAN X, SPANX, SPANX A, SPANX family member A, SPANX family, member A2, SPANXA, SPANXA1, SPANXA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
30014, 728712 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NS26 | |
SPANXA1, SPANXA2 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |