missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25000-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the Sp subfamily of Sp/XKLF transcription factors. Sp family proteins are sequence-specific DNA-binding proteins characterized by an amino-terminal trans-activation domain and three carboxy-terminal zinc finger motifs. This protein contains the least conserved DNA-binding domain within the Sp subfamily of proteins, and its DNA sequence specificity differs from the other Sp proteins. It localizes primarily within subnuclear foci associated with the nuclear matrix, and can activate or in some cases repress expression from different promoters.Especificaciones
SP2 | |
Polyclonal | |
Unconjugated | |
SP2 | |
KIAA0048, SP2, Sp2 transcription factor, Transcription factor Sp2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6668 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q02086 | |
SP2 | |
SP2 Fusion Protein Ag8060 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |