missing translation for 'onlineSavingsMsg'
Learn More

SP1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody

Marque:  Bio-Techne NB600-233FR

Informations supplémentaires : Peso : 0.00970kg

Code produit. 18215483

  • 620.93€ / 100 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock

Description

Description

SP1 Polyclonal specifically detects SP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification

Spécification

SP1
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Sp1 transcription factor, specificity protein 1, transcription factor Sp1, TSFP1
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
SP1
The immunogen recognized by this antibody maps to a region between residue 750 and the C-terminus (residue 785) using the numbering given in entry XP_028606.4 (GeneID 6667).
100 μL
Cancer, Phospho Specific, Transcription Factors and Regulators
6667
Store at 4C in the dark.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions