missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOLO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23911-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
SESTD1 was initially identified in mutant zebrafish with defects in the spontaneous contraction and touch response as a novel gene, solo, encoding a protein containing SEC14 and spectrin repeat domains. Other experiments indicated that SESTD1 interacts with the TRPC4 and TRPC5, members of the transient receptor potential channel family, via the TRPC calmodulin- and inositol 1, 4, 5-triphosphate receptor-binding domain and is essential for efficient receptor-mediated activation of TRPC5, suggesting that SESTD1 is a novel regulator of these TRPC proteins.Especificaciones
SOLO | |
Polyclonal | |
Unconjugated | |
SESTD1 | |
Protein Solo, SEC14 and spectrin domains 1, SESTD1, SOLO | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
228071, 295678, 91404 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80UK0, Q86VW0 | |
SESTD1 | |
SOLO Fusion Protein Ag21016 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |