Learn More
Brady™ Brass Padlocks
Product Code. 15693497 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Blue
Gray
Green
Red
Yellow
Unit Size:
Pack of 12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15693497 Gray Pack of 12
15603507 Black Pack of 12
15663497 Blue Pack of 12
15673497 Green Pack of 12
15653497 Red Pack of 12
15683497 Yellow Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15693497 Supplier Brady™ Supplier No. 051358
  • ONLINE EXCLUSIVE PRICE
    176.00€
    152.84€ / paquete de 12
    Ahorro 23.16€
    13% Descuento
Fecha estimada de envoi: 08-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Features solid brass body and shackle that resists corrosion and sparks.
  • 6-pin cylinder offers more unique key cuts for better tamper resistance
  • Locks are protected by coloured vinyl sheaths making it easy to color-code your lockout program
  • Padlocks are available keyed different
  • Lock labels sold separately
  • Two keys supplied with each padlock
Specifikationer
Product Type Brass Padlocks
Key Type Keyed Different
Alternate Part Number 51358
Color Gray
Material Brass
Description Brass Padlocks
Diameter (English) Shackle 0.25 in.
Diameter (Metric) Shackle 6.5 mm
Dimensions (L x W) 43.00 x 60.00 mm
For Use With (Application) Lockout Tagout
Height (Metric) 21 mm
Height (Metric) Shackle 21.00 mm
Material (Shackle) Brass
No. of Pins 6
Quantity 12 Pk.
Width (Metric) 33.5 mm
Width (Metric) Shackle 33.50 mm
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.