Learn More
Thermo Scientific™ SOLAμ™ SPE Plates
Product Code. 31174685 Shop All Thermo Scientific Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Includes:
HRP 2 mg/1 mL 96 well fixed plate
HRP 2 mg/1 mL 96 well removable plate
SAX 2 mg/1 mL 96 well fixed plate
SAX 2 mg/1 mL 96 well removable plate
SCX 2 mg/1 mL 96 well fixed plate
SCX 2 mg/1 mL 96 well removable plate
WAX 2 mg/1 mL 96 well fixed plate
WAX 2 mg/1 mL 96 well removable plate
WCX 2 mg/1 mL 96 well fixed plate
WCX 2 mg/1 mL 96 well removable plate
missing translation for 'unitSize':
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes missing translation for 'unitSize'
31174685 HRP 2 mg/1 mL 96 well removable plate -
15261808 HRP 2 mg/1 mL 96 well fixed plate Each
15271808 SCX 2 mg/1 mL 96 well fixed plate Each
15281808 SAX 2 mg/1 mL 96 well fixed plate Each
15291808 WCX 2 mg/1 mL 96 well fixed plate Each
15201818 WAX 2 mg/1 mL 96 well fixed plate Each
31174684 SCX 2 mg/1 mL 96 well removable plate -
31174686 SAX 2 mg/1 mL 96 well removable plate -
31174683 WCX 2 mg/1 mL 96 well removable plate -
31174687 WAX 2 mg/1 mL 96 well removable plate -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 31174685 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 60209001RW
 Restricted Item
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Remove variability and achieve highly consistent results. High reproducibility of Thermo Scientific™ SOLAμ™ SPE plates deliver greater sample success at low elution volumes.
Remove variability and optimize high throughput laboratory workflows. Thermo Scientific™ SOLAμ™Solid Phase Extraction (SPE) Well Plates are designed for bioanalytical and clinical research analysts who require cleaner, highly reproducible, and robust sample extraction using very low sample and solvent volumes in high throughput workflows. SOLAμ Well Plates achieve this due to the unique and award winning innovative frit-less SPE technology.
Ideal for Bioanalytical and Clinical Research Laboratories
Bioanalytical laboratories continue to drive to higher efficacy drugs and long acting formulations which require lower quantification limits. There is also the desire to take advantage of the replacement, refinement and reduction policy, which is resulting in reduced animal model use. The growth of biopharmaceuticals also brings into consideration additional analytical issues such as solvation and non-specific binding.
Meet Demanding Sample Preparation Requirements
  • Processing of low sample volumes
  • High sensitivity due to low elution volumes
  • High reproducibility
  • Low analytical sample failures
  • Ease of use
  • High throughput processing
SOLAμ is the first micro-elution product to meet the requirements of the bioanalyst in combination with a robust, easy to use format.
TRUSTED_SUSTAINABILITY
Specifications
Bed Weight 2 mg
For Use With (Application) Bioanalytical Applications - Hydrophobic Retention of Neutral Compounds with Complementary Retention of Polar Analytes. An All-Purpose Phase.
Product Line SOLAμ
Sorbent HRP (Reversed Phase Polymeric)
Type 96-well SPE Plate
Volume (Metric) Well 1 mL
Includes HRP 2 mg/1 mL 96 well removable plate
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.