Learn More
Sodium Tricyanomethanide, TRC
Product Code. 30006461
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006461 100 mg 100mg
30135853 500 mg 500mg
30138044 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006461 Supplier Toronto Research Chemicals Supplier No. TRCS673658100MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36603-80-2
Molecular Formula C4 N3 . Na
Molecular Weight (g/mol) 113.053
IUPAC Name sodium;methanetricarbonitrile
SMILES [Na+].N#C[C-](C#N)C#N
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C4N3.Na/c5-1-4(2-6)3-7;/q-1;+1
Formula Weight 112.999
Chemical Name or Material Sodium Tricyanomethanide