missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate Decahydrate, Honeywell Fluka™

Catalog Number p-7071893
Click to view available options
Cantidad:
100 g
2.5 kg
Este artículo no se puede devolver. Vea la política de devoluciones
13472-36-1
H20Na4O17P2
446.05
MFCD00149200
VZWGHDYJGOMEKT-UHFFFAOYSA-J
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
3084150
tetrasodium;phosphonato phosphate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
Este artículo no se puede devolver. Vea la política de devoluciones
13472-36-1
H20Na4O17P2
446.05
MFCD00149200
VZWGHDYJGOMEKT-UHFFFAOYSA-J
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
3084150
tetrasodium;phosphonato phosphate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Sodium Pyrophosphate Decahydrate
Cantidad 100 g
Fórmula lineal Na4P2O7 · 10H2O
Número UN NONH for all modes of transport
Formula Weight (peso de la fórmula) 446.06g/mol
Porcentaje de pureza ≥99%
Grado ACS Reagent
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'