Learn More
Sodium N-(4-Hydroxyphenyl)acetamide-2,2,2-d3 Sulfate, TRC
Product Code. 30328356
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328356 5 mg 5mg
30318090 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30328356 Supplier Toronto Research Chemicals Supplier No. TRCA1612295MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Acetaminophen-d3 Sulfate
CAS 1020718-78-8
Molecular Formula C8 2H3 H6 N O5 S
Quantity 5 mg
Synonym Acetamide-2,2,2-d3, N-[4-(sulfooxy)phenyl]- (ACI), N-[4-(Sulfooxy)phenyl]acetamide-2,2,2-d3 (ACI), 4-Acetaminophen-d3 sulfate, Acetaminophen sulfate-d3, N-(Acetyl-d3)-4-aminophenol O-sulfate, Paracetamol O-sulfate D3 (acetyl-D3), Paracetamol-d3 sulfate
InChI Formula InChI=1S/C8H9NO5S/c1-6(10)9-7-2-4-8(5-3-7)14-15(11,12)13/h2-5H,1H3,(H,9,10)(H,11,12,13)/i1D3
SMILES [2H]C([2H])([2H])C(=O)Nc1ccc(OS(=O)(=O)O)cc1
IUPAC Name [4-[(2,2,2-trideuterioacetyl)amino]phenyl] hydrogen sulfate
Molecular Weight (g/mol) 234.24
Formula Weight 234.039