missing translation for 'onlineSavingsMsg'
Learn More

Metatungstato de sodio monohidrato, Thermo Scientific Chemicals

Catalog Number 11318057
Click to view available options
Cantidad:
50 g
250 g
4 x 250 g
12141-67-2
Na6O39W12·H2O
MFCD00061466
Sodium tungstate (meta), Sodium polytungstate
12141-67-2
Na6O39W12·H2O
MFCD00061466
Sodium tungstate (meta), Sodium polytungstate

Sodium metatungstate monohydrate plays a vital role in sink and swim analysis. It is a derivative of sodium tungstate, which is a potential therapy for obesity. It acts as an inhibitor, which inhibits nucleoside-triphospahte- diphosphohydrolases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Sodium metatungstate monohydrate
Color Blanco
Forma física Polvo
Cantidad 4 x 250 g
Información de solubilidad Soluble in water.
Formula Weight (peso de la fórmula) 2986.13 (2968.12 Anhydrous)
Olor Odorless
Aspecto White powder
Densidad 5.47

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'