missing translation for 'onlineSavingsMsg'
Learn More

Solución solución volumétrica estándar Acculute de hidróxido de sodio, concentración final 0,5 N, Thermo Scientific Chemicals

Catalog Number p-7080420
Click to view available options
Cantidad:
1 U
6 U
Este artículo no se puede devolver. Vea la política de devoluciones
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodio;hidróxido
[OH-].[Na+]
Este artículo no se puede devolver. Vea la política de devoluciones
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodio;hidróxido
[OH-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Sodium hydroxide Acculute Standard Volumetric Solution
Nombre de nota Final Concentration 0.5N
Número UN UN1824
Formula Weight (peso de la fórmula) 40
Olor Odorless
Cantidad 1 U
Forma física Líquido

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'