Learn More
Sodium Ethyl Sulfate, TRC
Product Code. 30413959
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity Unit Size
30413959 2.5 g 2.5g
1 options
This item is not returnable. View return policy
Product Code. 30413959 Supplier Toronto Research Chemicals Supplier No. TRCS6253002.5G
 Restricted Item
Fecha estimada de envoi: 22-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl sulfate sodium salt (as ethyl sulfate)
CAS 546-74-7
Purity Grade Notes HPLC
Molecular Formula C2 H5 O4 S . Na
Quantity 2.5 g
Synonym Sulfuric acid','monoethyl ester','sodium salt (1:1), Ethyl sodium sulfate (6CI,7CI), Sulfuric acid','monoethyl ester','sodium salt (8CI,9CI), Sodium ethyl sulfate, Sodium sulfovinate, Ethyl Sulfate Sodium Salt
InChI Formula InChI=1S/C2H6O4S.Na/c1-2-6-7(3,4)5;/h2H2,1H3,(H,3,4,5);/q;+1/p-1
SMILES [Na+].CCOS(=O)(=O)[O-]
IUPAC Name sodium;ethyl sulfate
Molecular Weight (g/mol) 148.11
Formula Weight 147.9806
Percent Purity >95