missing translation for 'onlineSavingsMsg'
Learn More

Acetato de sodio, solución acuosa 3 M, pH 4,5, en autoclave, Thermo Scientific Chemicals

Catalog Number p-7042080
Click to view available options
Cantidad:
125 mL
250 mL
Este artículo no se puede devolver. Vea la política de devoluciones
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
[Na+].CC([O-])=O
Este artículo no se puede devolver. Vea la política de devoluciones
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
[Na+].CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Sodium acetate
Nombre de nota autoclaved
Fórmula lineal NaOOCCH3
Concentración 3M aq. soln.
Nombre IUPAC sodium acetate
Formula Weight (peso de la fórmula) 82.03
Olor Odorless
pH 4.5
Cantidad 125 mL
Forma física Líquida

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'