missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hidrato de 1-octaneoulfonato de sodio, grado HPLC, + 99 %, Thermo Scientific Chemicals
Click to view available options
Cantidad:
5 g
25 g
100 g
Descripción
Ion-pair reagent for HPLC analysisSodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Sodium 1-octanesulfonate hydrate |
Punto de fusión | >300°C |
Cantidad | 5 g |
Información de solubilidad | Soluble in water producing a clear and colorless solution. |
Formula Weight (peso de la fórmula) | 216.28 (Anhydrous) |
Porcentaje de pureza | ≥99% |
Grado | HPLC |
Forma física | cristalino/a/s |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'