missing translation for 'onlineSavingsMsg'
Learn More

SOD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

140.00€ - 399.00€

Especificaciones

Antígeno SOD2
Concentración 0.38 mg/mL
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16842685
Ver documentos Detalles de la promoción
Proteintech
24127-1-AP-20UL
20 μL
140.00€
20 microlitros
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 17-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16832685
Ver documentos Detalles de la promoción
Proteintech
24127-1-AP-150UL
150 μL
399.00€
150 microlitros
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 17-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Superoxide dismutase (SOD2) is an antioxidant enzyme involved in the defense system against reactive oxygen species (ROS). SOD catalyzes the dismutation reaction of superoxide radical anion (O2) to hydrogen peroxide, which is then catalyzed to innocuous O2 and H2O by glutathione peroxidase and catalase. Several classes of SOD have been identified. These include intracellular copper, zinc SOD (Cu, Zn-SOD/SOD-1), mitochondrial manganese SOD (Mn-SOD/SOD-2) and extracellular Cu, Zn-SOD (EC-SOD/SOD-3). SOD1 is found in all eukaryotic species as a homodimeric 32 kDa enzyme containing one each of Cu and Zn ion per subunit. The manganese containing 80 kDa tetrameric enzyme SOD2, is located in the mitochondrial matrix in close proximity to a primary endogenous source of superoxide, the mitochondrial respiratory chain. SOD3 is a heparin-binding multimer of disulfide-linked dimers, primarily expressed in human lungs, vessel walls and airways. SOD4 is a copper chaperone for superoxide dismutase (CCS), which specifically delivers Cu to copper/zinc superoxide dismutase. CCS may activate copper/zinc superoxide dismutase through direct insertion of the Cu cofactor.
Especificaciones

Especificaciones

SOD2
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
P04179, P07895, P09671
20656, 24787, 6648
SOD2 Fusion Protein Ag21388
Primary
-20°C
SOD2
0.38 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
IPO B, Mn SOD, MNSOD, SOD2, SODM
SOD2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications