Learn More
SOD2/Mn-SOD Antibody (012), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca:  Novus Biologicals NBP2-90139AF405
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213518
  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
SOD2/Mn-SOD Monoclonal specifically detects SOD2/Mn-SOD in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
SOD2/Mn-SOD
Monoclonal
Alexa Fluor 405
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
12
Immunohistochemistry, Immunohistochemistry-Paraffin
EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial
This antibody was obtained from a rabbit immunized with purified, recombinant Human SOD2/Mn-SOD (P04179-1; Lys25-Lys222).
0.1 mL
Cancer, Diabetes Research, Neuroscience, Tumor Suppressors
6648
Store at 4°C in the dark.