Learn More
DWK Life Sciences™ Sockets for Soda-lime Glass Spirit Lamps
Product Code. 11756978 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Aluminum
Unit Size:
Pack of 50
Product Code. Material Unit Size
11756978 Aluminum Pack of 50
1 options
This item is not returnable. View return policy
Product Code. 11756978 Supplier DWK Life Sciences™ Supplier No. 294030008
  • ONLINE EXCLUSIVE PRICE
    80.40€
    70.66€ / paquete de 50
    Ahorro 9.74€
    12% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
The spirit lamp is an alternative to the gas-powered Bunsen burner and is also suitable for heating cooking glasses and the materials they contain.
In order for substances to react with each other, a low heat input is often required. The spirit burner is used classically and in areas where direct gas extraction is not possible. It is an alternative to the well-known Bunsen burner.
  • Without filling tubulature
  • The ground cap ensures a tight seal and quick suffocation of the flame
Specifications
Material Aluminum
Description Sockets for spirit lamps (of aluminum)
For Use With (Application) Heating; Use with Spritus
No. per Pack 50/Pk.