missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNCAIP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Especificaciones
Antígeno | SNCAIP |
---|---|
Concentración | 0.37 mg/mL |
Aplicaciones | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16817814
|
Proteintech
17818-1-AP-20UL |
20 μL |
140.00€
20 microlitros |
Fecha estimada de envío: 13-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16807814
|
Proteintech
17818-1-AP-150UL |
150 μL |
399.00€
150 microlitros |
Fecha estimada de envío: 13-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Parkinson is the second most common neurodegenerative disease after Alzheimer′s. About 1 percent of people over the age of 65 and 3 percent of people over the age of 75 are affected by the disease. The mutation is the most common cause of Parkinson disease identified to date. Synuclein alpha interacting protein (Synphilin-1) contains several protein-protein interaction domains and interacts with alpha synuclein in neurons. Mutations of SNCAIP have been linked to Parkinson disease. The amino acid sequence of synphilin-1 shows extensive homology with its human counterpart, especially in regions containing ankyrin-like motifs and the coiled-coil domain. Expression of mouse synphilin-1 in tissues is similar to its human counterpart. Synphilin-1 has an important role in the formation of aggregates and cytotoxicity in Parkinson disease and that Dorfin may be involved in the pathogenic process by ubiquitylation of synphilin-1. Role of synphilin-1 in synaptic function and protein degradation and in the molecular mechanisms leading to neurodegeneration in Parkinson disease.Especificaciones
SNCAIP | |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q99ME3, Q9Y6H5 | |
67847, 9627 | |
SNCAIP Fusion Protein Ag12147 | |
Primary | |
-20°C | |
SNCAIP |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SNCAIP, Sph1, Synphilin 1, SYPH1 | |
Sncaip | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |