Learn More
Motic™ SMZ161 Microscope
Convincing optical performance, a small footprint, and robust mechanics makes this the ideal stereo zoom microscope for teaching situations. Motic™ SMZ161 Microscope is constructed with light, ESD-material components.
Marca:  Motic™ 1100201300022
Detalles adicionales : Peso : 1.20000kg
 Ver más versiones de este producto
Código de producto. 15495412
  • 636.09€ / cada uno
Fecha de envío estimada
del proveedor 22-11-2019
Añadir a la cesta
Descripción
Descripción
  • Greenough optical system.
  • Sharp, three-dimensioned images and wide field of view.
  • Interpupillary distance: 50-75mm.
  • Ergonomically designed for long hours of viewing without discomfort.
  • Suitable for industrial applications, assembly and inspection of small parts or integrated circuits on PCB, LCD production line.
  • Coarse focusing system with tension adjustment.
Especificaciones
Especificaciones
Stereo zoom microscope
SMZ161-BP
0.75X - 4.5X
WF10X/20mm
None
Black and white stage plates and dust cover
Pole-type stand without illumination, 280x220mm
Zoom system, ratio 1:6
Binocular head with diopter adjustment on both eyepiece tubes
1:6