missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMOX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Especificaciones
Antígeno | SMOX |
---|---|
Concentración | 0.47 mg/mL |
Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16824034
|
Proteintech
15052-1-AP-20UL |
20 μL |
140.00€
20 microlitros |
Fecha estimada de envío: 08-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16814034
|
Proteintech
15052-1-AP-150UL |
150 μL |
399.00€
150 microlitros |
Fecha estimada de envío: 08-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Polyamines are ubiquitous polycationic alkylamines which include spermine, spermidine, putrescine, and agmatine. These molecules participate in a broad range of cellular functions which include cell cycle modulation, scavenging reactive oxygen species, and the control of gene expression. These molecules also play important roles in neurotransmission through their regulation of cell-surface receptor activity, involvement in intracellular signalling pathways, and their putative roles as neurotransmitters. This gene encodes an FAD-containing enzyme that catalyzes the oxidation of spermine to spermadine and secondarily produces hydrogen peroxide. Multiple transcript variants encoding different isoenzymes have been identified for this gene, some of which have failed to demonstrate significant oxidase activity on natural polyamine substrates.Especificaciones
SMOX | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q99K82, Q9NWM0 | |
228608, 308652, 54498 | |
SMOX Fusion Protein Ag7035 | |
Primary | |
-20°C | |
SMOX |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C20orf16, PAO, PAO 1, PAOh1, Polyamine oxidase 1, SMO, SMOX, spermine oxidase | |
SMOX | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |