missing translation for 'onlineSavingsMsg'
Learn More

SMN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11708-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818973

  • 109.00€ / 20 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody, 11708-1-AP, raised against the recombit full-length human SMN2 protein, recognizes all isoforms of SMN protein.

The StARD (steroidogenic acute regulatory protein-related lipid transfer (START) domain containing) family of proteins is comprised of fifteen different members. All members contain the characteristic START domain and are believed to play key roles in the metabolism and transport of lipids. The StARD proteins are grouped into six subfamilies based on their START domain sequences. PC-TP (StARD2), StARD7, StARD10 and GPBP (StARD11) constitute one subfamily, namely the STARD2/PCTP group. StARD10, also known as PCTP2, PCTPL, NY-CO-28, CGI-52 or SDCCAG28, is widely expressed and functions in phospholipid transfer, binding to phosphatidylcholine and phosphatidylethanolamine. StARD10 can be found in sperm flagellum, potentially functioning as an enzyme involved in energy metabolism, and its expression is developmentally regulated in testis and mammary glands. StARD10 activity can be inhibited via phosphorylation by casein kinase II.
Especificaciones

Especificaciones

SMN
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35876, P97801, Q16637
C BCD541, Component of gems 1, Gemin 1, SMN, SMN1, SMN1,SMN, SMN2, SMNC, SMNT, Survival motor neuron protein
Rabbit
IgG
20 μL
-20°C
Primary
20595, 64301, 6606
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
SMN1
Liquid
Smn1
SMN Fusion Protein Ag2260
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documentos
Certificados