Learn More
Thermo Scientific™ Smart-Vue™ Alert
Replaces the use of an analog USB model for issuing Smart-Vue wireless monitoring alerts via voice and/or SMS.
Marca:  Thermo Scientific™ SMARTVUEALERT100
Detalles adicionales : Peso : 0.80000kg
 Ver más versiones de este producto
Código de producto. 15935170
  • 3011.00€ / cada uno
Fecha estimada de envío
13-03-2020
Añadir a la cesta
Especificaciones
Especificaciones
Smart-Vue Alert, 51-100 Sensors
Smart-Vue Alert
36 month software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors (dual modules count as 2 sensors). Requires SmartVueServer/SmartVue Client version 2.2 and above.
Voice and SMS
Wireless Monitoring