Learn More
Thermo Scientific™ Smart-Vue Pro Quatro Modules 868Mhz
Product Code. 17501076 Shop All Thermo Scientific Products
Change view
Click to view available options
Display Type:
LCD
Unit Size:
Each
Product Code. Display Type Unit Size
17501076 LCD Each
1 options
This item is not returnable. View return policy
Product Code. 17501076 Supplier Thermo Scientific™ Supplier No. SVPHWRMOD024
  • ONLINE EXCLUSIVE PRICE
    790.00€
    764.00€ / paquete de 1
    Ahorro 26.00€
    3% Descuento
View all promotions
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Increase efficiency with multi-sensor wireless monitoring
Smart-Vue Pro Quattro 868Mhz Only. Delivered with two 3.6V lithium batteries, two sensor mounting kit, one mounting holder, one antenna and two 1.5 m/5 ft. flat cable.
  • Bright 2.4 in. color touch screen with multiple views
  • Alarms locally with real -time audio-visual alarms on the modules
  • Audio visual alarms for local notification
  • Communication locally via Bluetooth and to the system via LoRaLow
  • AC power consumption through USB port access
  • Optional replaceable battery power with extended life (minimum of 1 year) allows continuous monitoring
  • Choice of wired and wireless sensors with automatic sensor detection
  • User traceability with customized levels of user access with encrypted password protection
Specifications
Type Module
Display Type LCD
Battery Type 3.6 V lithium battery
Battery Life 2 Years
Includes Two 3.6 V lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables
Certifications/Compliance Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance